missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF
Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.
Brand: Cytiva 6705-7500
This item is not returnable.
View return policy
Description
- Three-layer filter permits filtration of larger volumes
- Integrity testable
- With inlet vent
- Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0μm polysulfone
Serum Prefilter Capsules. Retention (with air purge), <2mL
For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.
Specifications
Autoclavable | |
Glass Microfiber, Polyethersulfone | |
535 cm2 | |
Polypropylene (PP) | |
Stepped Barb |
Polycap SPF 75 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 pc) | |
1 μm | |
For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions. | |
4.1 bar (60 psi) |