missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF

Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.

Brand:  Cytiva 2820

 View more versions of this product

Product Code. 11379134

  • 927.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Three-layer filter permits filtration of larger volumes
  • Integrity testable
  • With inlet vent
  • Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0 μm polysulfone

Serum Prefilter Capsules. Retention (with air purge), < 2 mL

For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Prefilter: Two Layers of Glass Microfiber
1 μm
For Serum Prefiltration. Handle Tissue Culture Media, Nutrients, Immunologicals, Enzymes, Diagnostic Standards/Controls, Biologicals, Buffers, Virus Suspensions.
1/2 in.
4.1 bar (60 psi)
Serum Prefilter
Polycap SPF 150 Capsule Filter, 1.0 μm, with SB inlet and outlet (1 Pc.)
Glass Microfiber, Polyethersulfone
1100 cm2
Polypropylene
Stepped Barb
1/2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers