Learn More
Cytiva Whatman™ Polycap™ HD Disposable Capsules: 75 HD
Offers high filtration efficiency and excellent filtrate purity due to its materials and methods of manufacture. Whatman™ Polycap™ HD Disposable Capsules fit between gross filters and microporous membrane filters used for final filtration.
97.58 € - 554.00 €
Specifications
Autoclavable Autoclavable
Material (Membrane) Monofilament Anisotropic Polypropylene
Filtration Area 805 cm2
For Use With (Application) Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. Offer broad capacity between gross paper filters and microporous membranes.
Housing Material Polypropylene (PP)
View More Specs
Products 12
Product Code Brand Pore Size Inlet Style Outlet Style Price Quantity & Availability  
Product Code Brand Pore Size Inlet Style Outlet Style Price Quantity & Availability  
11379124
View Documents
Cytiva
2712T
5 μm Female national pipe thread (FNPT) Female national pipe thread (FNPT)
450.00 €
Pack of 5
11319134
View Documents
Cytiva
2714
20 μm Hose barb (HB) Hose barb (HB)
486.00 €
Pack of 5
11309134
View Documents
Cytiva
2714T
20 μm Female national pipe thread (FNPT) Female national pipe thread (FNPT)
462.00 €
Pack of 5
10654282
View Documents
Cytiva
6703-7521
20 μm Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing
97.58 €
Each
11394604
View Documents
Cytiva
6703-7550
5 μm Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing
97.58 €
Each
11389124
View Documents
Cytiva
2712M
5 μm Male national pipe thread (MNPT) Male national pipe thread (MNPT)
450.00 €
Pack of 5
11339134
View Documents
Cytiva
2713T
10 μm Female national pipe thread (FNPT) Female national pipe thread (FNPT)
554.00 €
Pack of 5
11359134
View Documents
Cytiva
2713
10 μm Hose barb (HB) Stepped Barb (SB)
455.00 €
Pack of 5
11374604
View Documents
Cytiva
6703-7511
10 μm Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing
97.58 €
Each
11364604
View Documents
Cytiva
6703-7510
1 μm Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing Stepped barb for 10 to 12 mm (3/8 to 1/2 in.) tubing
97.58 €
Each
11369124
View Documents
Cytiva
2711T
1.0 μm Female national pipe thread (FNPT) Female national pipe thread (FNPT)
463.00 €
Pack of 5
11359124
View Documents
Cytiva
2710
0.45 μm Hose barb (HB) Hose barb (HB)
442.00 €
Pack of 5
Description
Description
  • Integrity testable
  • With inlet vent
  • Filter Media: Monofilament anisotropic polypropylene (MAPP) filter
  • Able to be sterilized by autoclaving with steam or ethylene oxide (EtO)
  • Retention (with air purge), <2mL
  • Max pressure: 4.1 bar (60 psi)
Filtration of aqueous solutions, biologicals, inks and pigments, photographic emulsions, food and beverages, solvents. Offer broad capacity between gross paper filters and microporous membranes.
Specifications
Specifications
Autoclavable
805 cm2
Polypropylene (PP)
Monofilament Anisotropic Polypropylene
Filtration of high-debris solutions and prefiltration of aggressive chemical solutions. Offer broad capacity between gross paper filters and microporous membranes.