missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ SG81 Grade Chromatography Paper
SG81 ion exchange sheets is a 0.27 mm combining cellulose and large pore silica gel for separations in which both partition and adsorption are important.
Brand: Cytiva 3668-915
This item is not returnable.
View return policy
Description
- Simultaneous development of multiple samples on the same sheet under identical conditions.
- Sequential development of the same samples with different solvents and/or different concentrations of the same solvent.
- Suitable for two-dimensional chromatography (change in direction of the solvent front) with possible improved resolution.
- Used in enzyme assays to separate product from reactant by charge or polarity.

Specifications
0.27 mm | |
Sheet | |
Cellulose, Large Pore Silica | |
46 cm | |
57 cm |
For Enzyme Assays to Separate Product From Reactant by Charge or Polarity | |
SG81 | |
18.1 in. | |
110 mm/30 min. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Whatman™ SG81 Grade Chromatography Paper > Flow rate: 110mm/30 min.; Grade SG81; Type: sheet; 18.11 x 22.4 in. (460 x 570mm)
Spot an opportunity for improvement?Share a Content Correction