missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Sterile Mixed Cellulose Ester Membranes, ME 25

Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium

Brand:  Cytiva 10407970


 View more versions of this product

Product Code. 11345384

  • 201.00 € / Pack of 100
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
White
ME 25
Sterile
ME 25
Hydrophilic
Medium
Grid
130°C
Membrane
1.85 in.
4.3 to 5.0 mg/cm2
Mixed Cellulose Ester
0.45 μm
To quantitatively evaluate microorganism counts
White
Without interleaving paper and pad
0.31cm
266°F
145 μm
12.5 sec./100mL/12.5cm2
47 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers