Learn More
Cytiva Whatman™ Sterile Mixed Cellulose Ester Membranes, ME 25
Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium
96.29 € - 874.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Basis Weight | 4.3 to 5.0 mg/cm2 |
Material | Mixed Cellulose Ester |
Pore Size | 0.45 μm |
Sterility | Sterile |
Product Code | Brand | Color | Diameter (Metric) | Grid Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) | Grid Color | Price | Quantity & Availability | |||
11355394
|
Cytiva
10409772 |
Black | 50 mm | White |
185.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11335314
|
Cytiva
10401670 |
White | 47 mm | Not Applicable |
165.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11345394
|
Cytiva
10409771 |
Black | 47 mm | White |
874.00 €
Pack of 1000 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||
11335394
|
Cytiva
10409770 |
Black | 47 mm | White |
183.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
11345384
|
Cytiva
10407970 |
White | 47 mm | White |
207.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
13247119
|
Cytiva
10406802 |
White | 50 mm | Black | N/A | ||||
13267119
|
Cytiva
10406801 |
White | 50 mm | Black | N/A | ||||
13237119
|
Cytiva
10406800 |
White | 47 mm | Black | N/A | ||||
13257119
|
Cytiva
10406803 |
White | 47 mm | Black | N/A | ||||
Description
Cytiva Whatman Sterile Mixed Cellulose Ester Membranes, ME 25, Black, Diameter Outside: 47mm, 1.85 in., Grid Color: White, Autoclavable: Yes, Grade: ME 25, Includes: With interleaving paper and without pad, Membrane Type: WME, Pore Size: 0.45um, Protein Binding Capacity: Medium
Specifications
Autoclavable | |
Mixed Cellulose Ester | |
Sterile | |
130°C |
4.3 to 5.0 mg/cm2 | |
0.45 μm | |
Medium | |
Membrane |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.