missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ 25mm Filtration Assembly With No. 5 Stopper Connection With Stainless Steel Support
Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
Brand: DWK Life Sciences 419338
21.73 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Includes:
- 316 stainless steel support with 120 mesh screen
- PTFE support screen gasket
- Glass support base
- 15mL graduated funnel
- Anodized aluminum clamp
- No. 5 silicone stopper
Description
- For filtration of viscous or proteinaceous solutions to produce ultra clean filtrate
- Glass components manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
25mm filtration assembly | |
PTFE | |
6/Cs. | |
Support Screen Gasket |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction