missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
GREENER_CHOICE

Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.

109.00 € - 130.00 €

Specifications

Material Aluminum
Quantity 1000/Cs.
Type Complete Tear-off Cap
View More Specs

Products 4
Product Code Brand Color Closure Size Price Quantity & Availability  
Product Code Brand Color Closure Size Price Quantity & Availability  
10633543
GREENER_CHOICE
View Documents
DWK Life Sciences
224193-01
Natural 20 mm
109.00 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
10326902
GREENER_CHOICE
View Documents
DWK Life Sciences
224193-05
Blue 20 mm
130.00 €
Pack of 1000
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
11718789
GREENER_CHOICE
View Documents
DWK Life Sciences
224192-07
Green 13 mm
125.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
10580463
GREENER_CHOICE
View Documents
DWK Life Sciences
224193-06
Red 20 mm
130.00 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
Description

Description

Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.

  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
Specifications

Specifications

Aluminum
1000/Cs.
Complete Tear-off Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.