Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Use as replacement caps or for vials that are provided without caps
Brand:  DWK Life Sciences 241317
177.49 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10042003
  • 196.00 € / Pack of 1000
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Screw Caps For Liquid Scintillation Vials
  • Screw caps for WHEATON liquid scintillation vials
  • Case of 1000 provides an economical alterative
  • Not autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cap Size: 22-400
Natural
Screw Cap
Liquid Scintillation Vial
Linerless
Polyethylene
22-400