missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask

Membrane cell culture flasks ideal for antibody and protein production

Brand:  DWK Life Sciences WCL1000-1

261.07 EUR valid until 2024-12-31
Use promo code "21939" to get your promotional price.


 View more versions of this product

Product Code. 12801803

  • 362.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: User manual with feeding and harvesting schedules
This item is not returnable. View return policy

Description

Description

High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.

  • Bulk media chamber allows for longer run times without media refreshing
  • Cell compartment concentrates cells, antibodies and proteins for simplified purification
  • For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.

02-912-745, 02-912-742

  • For adherent cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-745) or package three (02-912-742)
02-912-746, 02-912-743
  • For suspension cell cultivation
  • 350 mL media compartment
  • 5 mL cell compartment
  • Available as package of one (02-912-746) or package five (02-912-746)
02-912-744, 02-912-741
  • For suspension cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-744) or package three (02-912-741)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.527 oz.
15 mL
Suspension Cultures
1/Cs.
35.2 oz.
1000 mL
User manual with feeding and harvesting schedules
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers