missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket

Provide maximum surface between culture and glass flask

Brand:  DWK Life Sciences 356887

981.89 EUR valid until 2024-12-31
Use promo code "21939" to get your promotional price.


 View more versions of this product

Product Code. 10142691

  • 1145.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Provide maximum surface between culture and glass flask

  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silicone Rubber
4.5 cm
13.4 in.
Flask, impeller and caps
Autoclavable
10 to 44 cm
7 in.
3000 mL
34.1 cm
PTFE Resin
17.8 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.