Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Provide maximum surface between culture and glass flask
179.00 € - 1795.00 €
Specifications
Closure Material | Silicone Rubber |
---|---|
Surface Coating | PTFE Resin |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | ||||
10557131
|
DWK Life Sciences
356875 |
50 mL | Flask, impeller and caps |
362.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||
10694351
|
DWK Life Sciences
356879 |
250 mL | Flask, impeller and caps |
477.00 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
10556941
|
DWK Life Sciences
356873 |
25 mL | Flask, impeller and caps |
364.00 €
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
10345851
|
DWK Life Sciences
356890 |
8 L | Flask, impeller and caps |
1795.00 €
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
10142691
|
DWK Life Sciences
356887 |
3000 mL | Flask, impeller and caps |
1145.00 €
Each |
In Stock
Log in to see stock. |
||||
10374601
|
DWK Life Sciences
356884 |
1000 mL | Flask, impeller and caps |
785.00 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
10100721
|
DWK Life Sciences
356876 |
125 mL | Flask, impeller and caps |
440.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
10779721
|
DWK Life Sciences
356882 |
500 mL | Flask, impeller and caps |
682.00 €
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
12491421
|
DWK Life Sciences
356915 |
50 mL | Flask only |
179.00 €
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
Description
Provide maximum surface between culture and glass flask
- PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
- Silicone rubber-lined cap
- Autoclavable
Specifications
Silicone Rubber | |
PTFE Resin | |
Autoclavable |