Learn More
DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
127.00 € - 412.00 €
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass (Low Extractable) |
For Use With (Application) | Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage |
Thread Style | Screw |
Closure Material | Phenolic |
Product Code | Brand | Volume (Metric) | Closure Size | Material (Liner) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Closure Size | Material (Liner) | Price | Quantity & Availability | ||||
10242431
|
DWK Life Sciences
224881 |
2 mL | 8-425 | 14B Rubber |
228.00 €
Pack of 288 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
10739151
|
DWK Life Sciences
224882 |
4 mL | 13-425 | 14B Rubber |
133.00 €
Pack of 144 |
In Stock
Log in to see stock. ![]() |
||||
10160861
|
DWK Life Sciences
224884 |
8 mL | 15-425 | 14B Rubber |
209.00 €
Pack of 144 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||
11734159
|
DWK Life Sciences
224885 |
12 mL | 15-425 | 14B Rubber |
208.00 €
Pack of 144 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
10446151
|
DWK Life Sciences
225288 |
20 mL | 24-400 | 14B Rubber |
141.00 €
Pack of 72 |
In Stock
Log in to see stock. ![]() |
||||
10153372
|
DWK Life Sciences
225014 |
8 mL | 15-425 | 14B Rubber |
196.00 €
Pack of 144 |
Estimated Shipment: 14-11-2025
Log in to see stock. |
||||
10504291
|
DWK Life Sciences
224886 |
16 mL | 18-400 | 14B Rubber |
209.00 €
Pack of 144 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
11758769
|
DWK Life Sciences
225289 |
25 mL | 24-400 | 14B Rubber |
127.00 €
Pack of 72 |
In Stock
Log in to see stock. ![]() |
||||
10628135
|
DWK Life Sciences
225290 |
40 mL | 24-400 | 14B Rubber |
140.40 €
Pack of 72 |
In Stock
Log in to see stock. |
||||
11744159
|
DWK Life Sciences
224888 |
24 mL | 20-400 | 14B Rubber |
270.00 €
Pack of 144 |
In Stock
Log in to see stock. |
||||
10193242
|
DWK Life Sciences
W224582 |
4 mL | 13-425 | PTFE/14B Rubber |
186.00 €
Pack of 144 |
In Stock
Log in to see stock. |
||||
10376942
|
DWK Life Sciences
W224581 |
2 mL | 8-425 | PTFE/14B Rubber |
412.00 €
Pack of 288 |
In Stock
Log in to see stock. |
||||
10041332
|
DWK Life Sciences
W224584 |
8 mL | 15-425 | PTFE/14B Rubber |
226.00 €
Pack of 144 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||
11728779
|
DWK Life Sciences
W224585 |
12 mL | 15-425 | PTFE/14B Rubber |
250.00 €
Pack of 144 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||
11738779
|
DWK Life Sciences
W224586 |
16 mL | 18-400 | PTFE/14B Rubber |
295.00 €
Pack of 144 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||
11748779
|
DWK Life Sciences
W224588 |
24 mL | 20-400 | PTFE/14B Rubber |
321.00 €
Pack of 144 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
10215212
|
DWK Life Sciences
W224589 |
20 mL | 24-400 | PTFE/14B Rubber |
197.00 €
Pack of 72 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
10622373
|
DWK Life Sciences
W224590 |
25 mL | 24-400 | PTFE/14B Rubber |
212.00 €
Pack of 72 |
In Stock
Log in to see stock. |
||||
10463902
|
DWK Life Sciences
W224591 |
40 mL | 24-400 | PTFE/14B Rubber |
271.00 €
Pack of 72 |
In Stock
Log in to see stock. |
||||
10465961
|
DWK Life Sciences
225012 |
4.0 mL | 13-425 | 14B Rubber |
176.00 €
Pack of 144 |
In Stock
Log in to see stock. |
||||
Description
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
- Fabricated from glass tubing for uniform sidewall and bottom thickness
- Vials and caps are autoclavable
- Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
- Cap and Cap Liners
- Solid top, black phenolic caps
- 14B rubber-lined black phenolic screw cap for aqueous samples
- PTFE/14B rubber-lined black phenolic screw cap for organic samples
Order Info
Replacements caps may be purchased separately.
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements.
Specifications
Clear | |
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage | |
Phenolic | |
Screw Cap | |
M-T File™ and Vial Rack |
Borosilicate Glass (Low Extractable) | |
Screw | |
Autoclavable | |
ASTM Type I, Class A, USP Type I |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.