Learn More
DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
Brand:  DWK Life Sciences 224885
159.18 EUR valid until 2025-12-31
Use promo code "24350" to get your promotional price.
 View more versions of this product
Product Code. 11734159
  • 208.00 € / Pack of 144
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Cap and Cap Liners
    • Solid top, black phenolic caps
    • 14B rubber-lined black phenolic screw cap for aqueous samples
    • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Order Info
Replacements caps may be purchased separately.
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14B Rubber
19 mm
12 mL
Screw
Phenolic
Screw Cap
ASTM Type I, Class A, USP Type I
M-T File™ and Vial Rack
Clear
Borosilicate Glass (Low Extractable)
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
3.2 dr.
Autoclavable
68 mm
15-425
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached > With caps; 12mL; Clear; 14B Rubber; 15-425; 19 x 68mm