Learn More
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit
Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
7205.00 € - 17470.00 €
Specifications
Color Amber
Sterility Sterile
Disposable Single-use
For Use With (Application) Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging
Graduated No
View More Specs
Products 18
Product Code Brand Diameter (Metric) Outer Closure Color Closure Type Quantity Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Closure Color Closure Type Quantity Price Quantity & Availability  
17992661
View Documents
DWK Life Sciences
W991000A-CPMP
Blue Ultra Pure Serum Stopper 278/Pk.
15225.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17972661
View Documents
DWK Life Sciences
W991001A-CPMP
Blue Omni Flex Serum Stopper 278/Pk.
16670.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17952581
View Documents
DWK Life Sciences
W991002A-CPMP
Blue Omni Flex Igloo Lyo Stopper 278/Pk.
17365.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17902671
View Documents
DWK Life Sciences
W991003A-CPMP
Red Ultra Pure Serum Stopper 278/Pk.
15225.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17982581
View Documents
DWK Life Sciences
W991004A-CPMP
Red Omni Flex Serum Stopper 278/Pk.
16670.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17912671
View Documents
DWK Life Sciences
W991005A-CPMP
Red Omni Flex Igloo Lyo Stopper 278/Pk.
17470.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17942671
View Documents
DWK Life Sciences
W991006A-CPMP
22 mm Blue Ultra Pure Serum Stopper
7205.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17932671
View Documents
DWK Life Sciences
W991007A-CPMP
22 mm Blue Omni Flex Serum Stopper
8055.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17922671
View Documents
DWK Life Sciences
W991008A-CPMP
22 mm Blue Omni Flex Igloo Lyo Stopper
8065.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17952591
View Documents
DWK Life Sciences
W991009A-CPMP
22 mm Red Ultra Pure Serum Stopper
7250.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17972671
View Documents
DWK Life Sciences
W991010A-CPMP
22 mm Red Omni Flex Serum Stopper
8055.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17952601
View Documents
DWK Life Sciences
W991011A-CPMP
22 mm Red Omni Flex Igloo Lyo Stopper
8065.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17962671
View Documents
DWK Life Sciences
W991012A-CPMP
24 mm Blue Ultra Pure Serum Stopper 104/Pk.
7255.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17952671
View Documents
DWK Life Sciences
W991013A-CPMP
24 mm Blue Omni Flex Serum Stopper 104/Pk.
9225.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17982671
View Documents
DWK Life Sciences
W991014A-CPMP
24 mm Blue Omni Flex Igloo Lyo Stopper 104/Pk.
9460.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17992671
View Documents
DWK Life Sciences
W991015A-CPMP
24 mm Red Ultra Pure Serum Stopper 104/Pk.
7210.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17992571
View Documents
DWK Life Sciences
W991016A-CPMP
24 mm Red Omni Flex Serum Stopper 104/Pk.
9050.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
17912681
View Documents
DWK Life Sciences
W991017A-CPMP
24 mm Red Omni Flex Igloo Lyo Stopper 104/Pk.
9275.00 €
1 set
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
Description
Description
  • Six inventory matched kits conveniently packed into a single shipper case.
  • Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Focuses your effort on core competencies and reduces lead time
  • Streamlines process and frees up valuable clean room space USP /EP 2.9.19
  • Particulate Matter in Injections, USP /EP 2.6.1,
  • The Bacterial Endotoxins Test and USP / EP 2.6.14
  • The Sterility Test, USP /EP 3.2.1
  • Glass Containers for Pharmaceutical Use
  • USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time
  • Minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in amber for photosensitive contents
Specifications
Specifications
Amber
Single-use
No
CompletePAK ModPAK
Autoclavable
Serum Vial
Sterile
Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging
Vial, Stopper, and Seal
Bromobutyl
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit