Learn More
DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.
Brand:  DWK Life Sciences 125500
 View more versions of this product
Product Code. 12694773
  • 279.00 € / Pack of 6
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Connector
24-400, 24-400