Learn More
DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.
149.00 € - 279.00 €
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
12477371
|
DWK Life Sciences
W125495 |
13-425, 22-400 |
225.00 €
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
12694773
|
DWK Life Sciences
125500 |
24-400, 24-400 |
279.00 €
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
12495442
|
DWK Life Sciences
125499 |
20-400, 24-400 |
264.00 €
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
12465442
|
DWK Life Sciences
125494 |
13-425, 20-400 |
149.00 €
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
12475442
|
DWK Life Sciences
125496 |
13-425, 24-400 |
259.00 €
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
Description
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Phenolic connectors have a working temperature of up to 180°C
- Autoclavable