Learn More
DWK Life Sciences Wheaton™ Culture Vessel
Perform repeated wet or dry sterilization to these culture vessels, which are made of borosilicate glass
Brand:  DWK Life Sciences 348524
 View more versions of this product
Product Code. 12438952
  • 544.00 € / Pack of 4
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conforms to USP and ASTM requirements
  • 38, 45 or 51mm screw head
  • 2.6mm wall thickness
  • Bottle material made of borosilicate glass
  • Autoclavable
  • Reusable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
355 mm
110 mm