missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Culture Vessel

Perform repeated wet or dry sterilization to these culture vessels, which are made of borosilicate glass

544.00 € - 697.00 €

Specifications

Diameter (Metric) 110 mm
View More Specs

Products 2
Product Code Brand Height (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Price Quantity & Availability  
12438952
View Documents
DWK Life Sciences
348524
355 mm
544.00 €
Pack of 4
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
12448952
View Documents
DWK Life Sciences
348528
535 mm
697.00 €
Pack of 4
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
 
Description

Description

  • Conforms to USP and ASTM requirements
  • 38, 45 or 51mm screw head
  • 2.6mm wall thickness
  • Bottle material made of borosilicate glass
  • Autoclavable
  • Reusable

Specifications

Specifications

110 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.