Learn More
DWK Life Sciences Wheaton™ E-C Sample Vials
Economically priced by packaging vials and screw caps separately. Wheaton™ E-C Sample Vials are available in either clear or amber-colored glass.
Brand:  DWK Life Sciences 224746
313.47 EUR valid until 2024-12-31
Use promo code "21960" to get your promotional price.
 View more versions of this product
Product Code. 10550372
  • 349.00 € / Pack of 200
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Economically priced by packaging vials and screw caps separately. Wheaton™ E-C Sample Vials are available in either clear or amber-colored glass.
  • Clear vials made from low extractable borosilicate glass that provides superior chemical resistance
  • Clear vials conform to USP Type I and ASTM E 438 Type I, Class A requirements
  • Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products
  • Vials packed in shrink-wrapped packs of 100 which reduces corrugate contamination
  • 14B rubber lined black phenolic screw caps for aqueous samples
  • PTFE-faced 14B rubber lined black phenolic screw cap for organic samples
  • Vials and caps are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE/14B Rubber
21 mm
16 mL
4.3 dr.
Autoclavable
73 mm
13-425
Clear
Borosilicate Glass
Screw
Phenolic
Screw Cap
ASTM Type I, USP Type I, Class A