missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™

Vial File provides a compact, easily accessible means of sample storage

Brand:  DWK Life Sciences 224952

 View more versions of this product

Product Code. 10206112

  • 115.00 € / Pack of 40
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Vial File provides a compact, easily accessible means of sample storage

Caps attached to vials.

  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products

Compliance

Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.

TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

PTFE/Silicone
15 mm
4 mL
No
1.08 dr.
Autoclavable
48 mm
Clear
Borosilicate Glass (Low Extractable)
Used for storing and transporting vials
Screw
Phenolic
Screw Cap (Open-top)
ASTM E438, USP Type I, Type I, Class A
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ver productos accesorios
Ofertas especiales

Ofertas especiales