Learn More
DWK Life Sciences Wheaton™ Micro Tissue Grinders
Used in extremely precise work when delicate hand operation is required
161.00 € - 322.00 €
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass
Product Type Micro Disposable Tissue Grinder
For Use With (Application) Used in extremely precise work when delicate hand operation is required
View More Specs
Products 3
Product Code Brand Capacity Length (English) Tube Diameter (English) Outside Price Quantity & Availability  
Product Code Brand Capacity Length (English) Tube Diameter (English) Outside Price Quantity & Availability  
11531305
View Documents
DWK Life Sciences
357844
0.003 oz., 0.1mL 4.33 in. 0.15 in.
205.00 €
Pack of 2
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
10449651
View Documents
DWK Life Sciences
357848
0.006 oz., 0.2mL 4.52 in. 0.27 in.
161.00 €
Pack of 2
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
11551305
View Documents
DWK Life Sciences
357860
0.02 oz., 0.75mL 3.46 in. 0.31 in.
322.00 €
Pack of 25
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
Description
Description
Used in extremely precise work when delicate hand operation is required
  • Made from low extractable borosilicate glass
Potter-Elvehjem Micro Tissue Grinder
  • Reservoir and pouring lip provide easy decanting of micro sample
  • Unit can be autoclaved / sterilized to prevent cross contamination between samples
  • Pestle clearance range: 0.1 – 0.15mm
Micro Tissue Grinder with PTFE Spacer
  • PTFE spacer aligns the pestle and reduces aerosoling of the product
  • Includes a finely ground glass pestle and tube
  • Autoclavable
Disposable Micro Tissue Grinder
  • Disposable to eliminate cross contamination and useful for radioactive samples
  • Small grinder area allows for high sample recovery
Specifications
Specifications
Autoclavable
Borosilicate Glass
Micro Disposable Tissue Grinder
Used in extremely precise work when delicate hand operation is required