Learn More
DWK Life Sciences™ Wheaton™ Prescored Gold-Band™ Amber Ampules
Protect light-sensitive samples
Brand: DWK Life Sciences™ 176792
74.56 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Amber borosilicate glass
- Prescored to eliminate filing
- Gold-Band feature indicates prescoring
- Stems can be pull sealed or tip sealed
Compliance
Meet USP Type 1 and ASTM™ Type I, Class B standards

Specifications
1 mL | |
Prescored Ampule | |
1.05 cm | |
6.7 cm | |
USP Type 1, ASTM Type I, Class A |
Borosilicate Glass | |
Amber | |
0.413 in. | |
2.63 in. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.