Learn More
DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
Screw thread finish eliminates the need for crimping tools
Brand: DWK Life Sciences W224218
101.50 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with CTC Analytics Systems requiring magnetic closures
- Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
Specifications
PTFE, Silicone | |
White PTFE/Transparent Blue Silicone | |
Screw Cap |