Learn More
DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
Screw thread finish eliminates the need for crimping tools
112.00 €
Description
- For use with CTC Analytics Systems requiring magnetic closures
- Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
Specifications
Screw Cap |