missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Staining Jars and Accessories

Staining Jars and Accessories

Brand:  DWK Life Sciences 900470

 View more versions of this product

Product Code. 10264732

  • 282.00 € / Pack of 6
Estimated Shipment: 14-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Coplin Staining Jar:

  • Made from soda-lime glass
  • 60mL capacity
Polypropylene Slide Grip:
  • Allows for easy and safe transfer of five slides
  • Fits into Wheaton Coplin staining jars

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Closure Included
Round
Soda Lime Glass
Staining
5 slides vertically or 10 slides back to back
1.02 in.
1.02 in.
3.54 in.
9 cm
Glass
65 mL
Coplin Staining Jar
Wheaton coplin staining jars
1.02 x 1.02 in. (2.6 x 2.6 cm)
26 mm
26 mm
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers