Learn More
DWK Life Sciences Wheaton™ Staining Jars and Accessories
Staining Jars and Accessories
54.45 € - 293.00 €
Specifications
For Use With (Application) Staining
View More Specs
Products 2
Product Code Brand Material Slide Count Type Price Quantity & Availability  
Product Code Brand Material Slide Count Type Price Quantity & Availability  
10492172
View Documents Promotion Details
DWK Life Sciences
900575
Polypropylene 5 Slide Grip
54.45 €
Pack of 2
Estimated Shipment: 10-04-2025
to see stock.
Add to basket
10264732
View Documents Promotion Details
DWK Life Sciences
900470
Soda Lime Glass 5 slides vertically or 10 slides back to back Coplin Staining Jar
293.00 €
Pack of 6
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
Description
Description
Coplin Staining Jar:
  • Made from soda-lime glass
  • 60mL capacity
Polypropylene Slide Grip:
  • Allows for easy and safe transfer of five slides
  • Fits into Wheaton Coplin staining jars
Specifications
Specifications
Staining