Learn More
DWK Life Sciences Wheaton™ V Vial™ with Crimp Top Finish; no Closures
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
Brand:  DWK Life Sciences W986211NG
 View more versions of this product
Product Code. 12663355
  • 143.00 € / Pack of 12
Estimated Shipment: 30-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
  • Not graduated
  • Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
  • Conical interior bottoms are designed to provide total downward drainage for maximum sample retrieval
Order Info
Closures are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.196 in.
Clear
Borosilicate Glass (Low Extractable)
Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Conical V
Crimp Cap
ASTM E438, USP Type I, Type I, Class A
5 mm
12 mm
0.1 mL
No
0.027 dr.
32 mm