Learn More
Nasco Whirl-Pak™ Write-On Stand-Up Thio Sample Bag
GREENER_CHOICE
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
Brand:  Nasco B01402WA
42.87 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
 View more versions of this product
Product Code. 11757954
  • 50.00 € / Pack of 100
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
  • A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself.
  • No rack or other holder is needed to keep the bag upright.
  • Forget washing and sterilizing glass bottles and disposing of expensive vials.
  • Sterile and unbreakable
Specifications
Specifications
Stand-up Bag
118 mL
25 bags/pouch, 100 bags/box
18.5 x 7.5 cm (7.25 x 3 in.)
18.5 cm
7.5 cm
4 oz.
0.064 mm
2.50 mil
7.25 in.
3 in.