Learn More
Nasco Whirl-Pak™ Write-On Stand-Up Thio Sample Bag
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
50.00 € - 94.00 €
Product Code | Brand | Capacity (English) | Dimensions (L x W) | Thickness (English) | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Dimensions (L x W) | Thickness (English) | Capacity (Metric) | Price | Quantity & Availability | ||
11757954
|
Nasco
B01402WA |
4 oz. | 18.5 x 7.5 cm (7.25 x 3 in.) | 2.50 mil | 118 mL |
50.00 €
Pack of 100 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
11787954
|
Nasco
B01403WA |
10 oz. | 23 x 11.5 cm (9 x 4.5 in.) | 3 mil | 300 mL | N/A | |||
Description
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
- A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself.
- No rack or other holder is needed to keep the bag upright.
- Forget washing and sterilizing glass bottles and disposing of expensive vials.
- Sterile and unbreakable
Specifications
Stand-up Bag | |
25 bags/pouch, 100 bags/box |