missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ White Dosing Spoon
BIOPHARMAGREENER_CHOICE

Volumetric and dosing spoons can be used to precisely measure small amounts of powder, granules, and liquids.

Brand:  Buerkle™ 5378-4013

 View more versions of this product

Product Code. 16515230

  • 72.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
  • Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
  • The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed, and the food and beverage industry
  • Sterile versions are individually packaged
  • With stripper edge
  • For single use
  • Cleanroom manufactured (Class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Spoon
Dosing spoons with stripper edge for exact measuring
Single-use
White
Especially for The Food, Animal Feed and Pharmaceutical Industries
Polystyrene
10 mL
Sterile
Food Contact, Reach, RoHS
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number242709 Certificate TypeCertificate of Compliance Date26/07/2024 Catalog Number5378-4013
Lot Number232999 Certificate TypeCertificate of Compliance Date25/07/2023 Catalog Number5378-4013
Lot Number222603 Certificate TypeCertificate of Compliance Date27/07/2022 Catalog Number5378-4013
Lot Number215913 Certificate TypeCertificate of Compliance Date18/03/2022 Catalog Number5378-4013
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ White Dosing Spoon > Sterile; Capacity: 10 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.