missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ White Polyethylene Laminate Category I Protective Coveralls
81.20 € - 103.02 €
Specifications
Product Type | Protective Coveralls |
---|---|
Waist Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 2-panel |
Product Code | Brand | dimSize | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimSize | Price | Quantity & Availability | |||||
14880972
|
3M™
7000089610 |
||||||||
14850972
|
3M™
7000089606 |
||||||||
14830972
|
3M™
7000089605 |
||||||||
14810972
|
3M™
7000089604 |
||||||||
11913734
|
3M™
7000089607 |
||||||||
14820972
|
3M™
7000089608 |
||||||||
14840972
|
3M™
7000089607 |
||||||||
14870972
|
3M™
7000089609 |
||||||||
Description
Specifications
Protective Coveralls | |
Elastic | |
2-panel |
Elastic | |
Hooded | |
Category I |