missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Scott™ Control Interfold 1 Ply Paper Hand Towels 6689, 15 Packs
GREENER_CHOICE

Interfold hand towels made with Airflex Technology are soft, strong, and highly absorbent. Ideal for busy offices and hotel washrooms. European Ecolabel certified paper hand towels for environmental assurance.

Brand:  Kimberly-Clark™ 6689

69.09 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12678855

  • 126.00 € / Pack of 15
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

15 Packs x 304 White Paper Hand Towels (4,560 Total) Each pack of Scott Control interfold hand towels contains 304 small, 1 ply, white hand towels Made using unique Airflex Technology, every paper towel is soft, strong and highly absorbent even when wet, providing an ideal hand drying solution for busy offices and hotel washrooms The clever, interfolded design dispenses one paper hand towel at a time, improving workplace hygiene, reducing wastage and refill frequency and providing exceptional cost in use Compressed packs ensure up to 18% more hand towels per case and these disposable paper towels are also European Ecolabel certified for hand hygiene with environmental assurance Compatible with a variety of Kimberly-Clark Professional and Aquarius folded hand towel dispensers Elevate your washroom experience with trusted and familiar Scott Control washroom solutions, designed to support compliance control and help you stop the spread of germs in the workplace. For busy washrooms demanding consistent hand hygiene and efficiency, choose Scott Control interfold hand towels. Scott Control hand towels are softer, more absorbent and stronger than conventional towels, providing consistent quality at an affordable price. Unique Airflex technology ensures that each lightweight single-ply paper towel is absorbent and strong, even when wet, preventing them from becoming soggy or ripping. These small disposable hand towels are designed to be kind to the skin for frequent hand drying, actively encouraging hand hygiene compliance. Scott Control interfold hand towels are designed to dispense one paper hand towel at a time. This single sheet dispensing method is designed to improve hand hygiene by minimising the risk of contamination when hand drying as the user only touches the sheet they're using. Dispensing one paper towel at a time is also designed to reduce waste and refill frequency as the user only takes the hand towels they need, providing exceptional cost in use. Scott Control hand towels carry the European Ecolabel; your guarantee that they are manufactured responsibly at every step of the process. The compressed packs also ensure up to 18% more paper towels per case. Scott Control interfold hand towels are compatible with a variety of Kimberly-Clark paper towel dispenser options: Aquarius folded hand towel dispensers in white (product codes 6945 and 6956) or black (code 7171), a high capacity dispenser (code 9962) and a high image stainless steel dispenser (code 8971). Each 1 ply paper towel measures 21cm (L) x 20cm (W). 15 Packs x 304 White Paper Hand Towels (4,560 Total)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Interfolded Hand Towel
54 cm
40 cm
6.495 kg (14.289 lb.)
White
Interfolded sheets
Aquarius™ Folded Hand Towel Dispenser
1-ply
1.77 ft.
1.31 ft.
Paper
ISO 14001, ISO 9001
210 x 220 mm
Food Contact Areas, Offices, Hotel Washrooms
15 x 274 Sheets
Small
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Scott™ Control Interfold 1 Ply Paper Hand Towels 6689, 15 Packs > Dimensions (L x W): 210 x 220 mm, Color: White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.