missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WypAll™ X70 Cloth, White

Ideal for medium-duty maintenance. White WypAll X70 Cloth available as quarter folded wipers wrapped in a splash-proof, water-repellent polybag.

Brand:  Kimberly-Clark™ 7867

130.33 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12373790

  • 401.00 € / Pack of 6
Estimated Shipment: 06-05-2025
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A repeat-use wiper with HydroKnit technology
  • Highly absorbent, strong and durable (wet or dry)
  • Resists most solvents, reduces residue, allows fast cleaning and helps reduce costs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Medium-duty Industrial Wiper
380 mm
420 mm
White
For medium-duty maintenance; wiping away oil, grease, grime, solvents, cleaning tools, machinery, metal surfaces, wiping up cutting fluids protecting parts
6 Pk.
Non-sterile
15 in.
16.5 in.
Non-woven Polypropylene Cellulose
380 x 420 mm (15 x 16.5 in.)
Wypall wiper dispenser
1-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ WypAll™ X70 Cloth, White > Dimensions (L x W): 380 x 420 mm (15 x 16.5 in.), Packaging Quantity: 6 Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.