Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-488
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of polyester and is used for thermal transfer printers.
Brand:  Brady™ 029895
 View more versions of this product
Product Code. 15685604
  • 114.00 € / 1 roll
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in various sizes
Specifications
Specifications
UL Recognized; CSA Accepted
Recommended ribbon series: R4300; R6200
White
Roll
Thermal Transfer Printer Label
BBP™81 Label Printer, IP™ Series, PR Plus Series Printers
50.8 x 6.35 mm (W x H)
2 in.
0.25 in.
THT-15-488-2.5
Acrylic, Permanent
Matte
Polyester (B-488)
Asset Tracking, Barcode Labeling, Circuit Board Identification - Through Hole Top, Component and Equipment Identification, General and Industrial Identification, Rating Plates
Rectangle
0.0036 in.
50.8 mm
6.35 mm