Learn More
Wright Stain, pure, certified, Thermo Scientific Chemicals
28.64 € - 144.72 €
Chemical Identifiers
CAS | 68988-92-1 |
---|---|
Molecular Formula | C36H27Br4N3O5S+2 |
Molecular Weight (g/mol) | 933.304 |
MDL Number | MFCD00082143 |
InChI Key | AXIKDPDWFVPGOD-UHFFFAOYSA-O |
PubChem CID | 25113599 |
IUPAC Name | [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid |
SMILES | CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10537182
|
Thermo Scientific Acros
422640100 |
10 g | Glass bottle |
28.64 €
10g |
In Stock
Log in to see stock. |
||||
10637761
|
Thermo Scientific Acros
422640250 |
25 g | Glass bottle |
53.27 €
25g |
In Stock
Log in to see stock. |
||||
10219393
|
Thermo Scientific Acros
422641000 |
100 g | Glass bottle |
144.72 €
100g |
Estimated Shipment: 11-03-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68988-92-1 | |
933.304 | |
AXIKDPDWFVPGOD-UHFFFAOYSA-O | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid |
C36H27Br4N3O5S+2 | |
MFCD00082143 | |
25113599 | |
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O |
Specifications
68988-92-1 | |
MFCD00082143 | |
AXIKDPDWFVPGOD-UHFFFAOYSA-O | |
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O | |
933.304 | |
Pure | |
Green | |
Wright Stain, Pure |
C36H27Br4N3O5S+2 | |
Solubility in water: insoluble. | |
Authentic | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid | |
25113599 | |
Complies | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.