missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WypAll™ L10 Food and Hygiene Wiping Paper, 24 Rolls x 165 Paper Wipes
GREENER_CHOICE

Compact wiping paper is designed for light duty hygiene wiping and ideal for tasks like cleaning food preparation and service surfaces. The embossing technology delivers a great balance of performance and value. Compatible with WypAll™ small roll dispenser 7041. 24 rolls x 165 sheets, 1 ply (3,960 total)

Brand:  Kimberly-Clark™ 7236

138.25 EUR valid until 2025-12-31
Use promo code "24298" to get your promotional price.


 View more versions of this product

Product Code. 16388338

  • 262.00 € / 24 rolls
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • L10 compact wiping roll contains 24 rolls x 165 wiping paper sheets, 1 ply sheets (3,960 total)
  • Dry cleaning wipes designed for light duty hygiene wiping and perfect for tasks like cleaning food preparation and service surfaces
  • Embossing technology in the wiping paper gives wipes a great balance of performance and value
  • Compact roll format for portability and ease of use
  • Designed for use with the WypAll wiping paper dispenser (product code 7041)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wiping Paper
48.4 cm
36.3 cm
White
WypAll™ Small Roll Dispenser 7041
Non-sterile
14.17 in.
14.17 in
Paper
Compact Roll
1-ply
Recycled F2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Specification Sheets

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ WypAll™ L10 Food and Hygiene Wiping Paper, 24 Rolls x 165 Paper Wipes > Color: White

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.