Learn More
Thermo Scientific™ X-Gal
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
39.75 € - 158.00 €
Specifications
Product Type X-Gal
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10203430
View Documents
Thermo Scientific™
R0401
0.5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10365410
View Documents
Thermo Scientific™
R0404
1 g
83.00 €
1g
10490470
View Documents
Thermo Scientific™
R0402
2 x 1 g
158.00 €
2g
Description
Description
X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.
Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
Specifications
Specifications
X-Gal
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ X-Gal