missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ X-Gal

Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.

Brand:  Thermo Scientific™ R0404

58.04 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10365410

  • 83.00 € / 1g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.

Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorimetric
1 g
Beta-Gal Substrate, X-Gal
X-Gal
Chemical Substrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ X-Gal > 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.