Learn More
ABEBA™ X-Light Black Safety Trainers S1
ABEBA™ X-Light Black Safety Trainers S1 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
Brand:  ABEBA™ 7131038/37
 View more versions of this product
Product Code. 16378331
  • 114.00 € / 1 pair
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 joules)
  • Black smooth leather
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Specifications
CE EN ISO 20345:2012 S2, SRC, ESD
Black
Breathable lining with Silver ions
37
Polyurethane
Steel
CE EN ISO 20345:2012, DIN EN 61340
Laces
Textile
Leather
Slip and abrasion resistant, shock absorbing, ESD dissipative
Trainer
1 Pair
Water