Learn More
ABEBA™ X-Light Black Safety Trainers S1
ABEBA™ X-Light Black Safety Trainers S1 with silver ions lining to reduce odor and a replaceable Active Comfort insole.
114.00 €
Specifications
Certifications/Compliance CE EN ISO 20345:2012 S2, SRC, ESD
Closure Type Laces
Color Black
Eyelets Material Textile
Liner Material Breathable lining with Silver ions
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16358331
View Documents
ABEBA™
7131038/35
35
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16368331
View Documents
ABEBA™
7131038/36
36
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16378331
View Documents
ABEBA™
7131038/37
37
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16388331
View Documents
ABEBA™
7131038/38
38
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16398331
View Documents
ABEBA™
7131038/39
39
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16308341
View Documents
ABEBA™
7131038/40
40
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16318341
View Documents
ABEBA™
7131038/41
41
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16328341
View Documents
ABEBA™
7131038/42
42
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16338341
View Documents
ABEBA™
7131038/43
43
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16348341
View Documents
ABEBA™
7131038/44
44
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16358341
View Documents
ABEBA™
7131038/45
45
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16368341
View Documents
ABEBA™
7131038/46
46
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16378341
View Documents
ABEBA™
7131038/47
47
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
16388341
View Documents
ABEBA™
7131038/48
48
114.00 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 joules)
  • Black smooth leather
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Specifications
CE EN ISO 20345:2012 S2, SRC, ESD
Black
Breathable lining with Silver ions
Slip and abrasion resistant, shock absorbing, ESD dissipative
Trainer
1 Pair
Water
Laces
Textile
Leather
Polyurethane
Steel
CE EN ISO 20345:2012, DIN EN 61340