Learn More
Abeba™ X-Light Safety Loafer
Abeba X-Light White Safety Loafer - Smooth leather safety shoes with elastic instep.
Brand:  Abeba™ 711032/48
64.14 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17652045
  • 117.00 € / 1 pair
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
Elastic Band
Leather
Steel
White
48
CE, EN ISO 20345:2011, S2, SRC