missing translation for 'onlineSavingsMsg'
Learn More

Epredia X50 Slides ground 45°, ISO 8037/1 1,0 mm

Brand:  Epredia AB00000102E01MNZ10

3.50 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


X50 Slides ground 45°, ISO 8037/1 1,0 mm

Product Code. 16211551

  • 8.50 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia X50 Slides ground 45°, ISO 8037/1 1,0 mm >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.