missing translation for 'onlineSavingsMsg'
Learn More

UVP XX-Series UV Bench Lamp Replacement Tubes

UVP™ XX-Series UV Bench Lamp Replacement Tubes are interchangeable UV tubes available in shortwave, longwave and mid-range wavelengths.

Brand:  UVP 34-0067-01

 View more versions of this product

Product Code. 12726508

  • 87.45 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The tubes in lamp fixtures are interchangeable for applications that require different wavelengths.
  • Order tubes of the same wattage as the lamp
  • Tubes available in 254nm, 302nm and 365nm
  • Shortwave UV is appropriate for sterilization and sanitation 
  • Longwave/Midrange UV is appropriate for fluorescence and inspection
Specifications

Specifications

Replacement UV Tube
254 nm
Sterilization and sanitation
23 in.
1 Ea.
20 W
Replacement UV tube for XX-Series benchtop lamp
Model XX-20 bench lamp
610 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.