missing translation for 'onlineSavingsMsg'
Learn More

UVP XX-Series UV Bench Lamp Replacement Tubes

UVP™ XX-Series UV Bench Lamp Replacement Tubes are interchangeable UV tubes available in shortwave, longwave and mid-range wavelengths.

54.05 € - 95.20 €

Specifications

Product Type Replacement UV Tube
Description Replacement UV tube for XX-Series benchtop lamp
Packaging Quantity 1 Ea.
View More Specs

Products 4
Product Code Brand Wavelength Wattage For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Wavelength Wattage For Use With (Equipment) Price Quantity & Availability  
12726508
View Documents
UVP
34-0067-01
254 nm 20 W Model XX-20 bench lamp
87.45 €
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
12803858
View Documents
UVP
34-0067-02
254 nm 40 W Model XX-40S bench lamp
95.20 €
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
12803848
View Documents
UVP
34-0008-01
254 nm 15 W Model XX-15 bench lamp
54.05 €
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
11807374
View Documents
UVP
34-0039-01
302 nm 15 W Model XX-15 bench lamp
82.35 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
Description

Description

The tubes in lamp fixtures are interchangeable for applications that require different wavelengths.
  • Order tubes of the same wattage as the lamp
  • Tubes available in 254nm, 302nm and 365nm
  • Shortwave UV is appropriate for sterilization and sanitation 
  • Longwave/Midrange UV is appropriate for fluorescence and inspection
Specifications

Specifications

Replacement UV Tube
Replacement UV tube for XX-Series benchtop lamp
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.