missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast Extract

Used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.

24.52 € - 71.56 €

Specifications

Form Powder
Packaging Type Glass Bottle
Sensitivity Hygroscopic

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346849
View Documents Promotion Details
Thermo Scientific Chemicals
H26769.22
100 g
24.52 €
100g
In Stock
to see stock.
Add to basket
 
11356849
View Documents Promotion Details
Thermo Scientific Chemicals
H26769.36
500 g
71.56 €
500g
In Stock
to see stock.
Add to basket
 
Description

Description

Yeast extract is used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Powder
Glass Bottle
Hygroscopic
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-387-9

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only