missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

46.23 € - 804.00 €

Chemical Identifiers

CAS 13760-80-0
Molecular Formula F3Yb
Molecular Weight (g/mol) 230.04
MDL Number MFCD00049615
InChI Key XASAPYQVQBKMIN-UHFFFAOYSA-K
Synonym ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
PubChem CID 21694467
SMILES [F-].[F-].[F-].[Yb+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366218
SDS View Documents
Thermo Scientific Alfa Aesar
011189.03
1 g
46.23 €
1g
Please to purchase this item. Need a web account? Register with us today!
11356218
SDS View Documents
Thermo Scientific Alfa Aesar
011189.09
10 g
165.83 €
10g
Please to purchase this item. Need a web account? Register with us today!
11346218
SDS View Documents
Thermo Scientific Alfa Aesar
011189.18
50 g
804.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ytterbium(III) fluoride is used in the preparation and study of fluorinated glasses. Further, it is employed as a laboratory reagent and in optical glasses and structural ceramics. It finds application in catalysts, electrical components and photo-optical materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13760-80-0
230.04
XASAPYQVQBKMIN-UHFFFAOYSA-K
21694467
F3Yb
MFCD00049615
ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
[F-].[F-].[F-].[Yb+3]

Specifications

1157°C
Lump/Powder
MFCD00049615
ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
XASAPYQVQBKMIN-UHFFFAOYSA-K
ytterbium(3+) trifluoride
21694467
99.99%
Odorless
8.2 g/mL
Ytterbium(III) fluoride, Anhydrous
13760-80-0
F3Yb
UN3288
Slightly soluble in strong mineral acids. Insoluble in water.
[F-].[F-].[F-].[Yb+3]
230.04
230.04
Reacton™
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-354-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.