missing translation for 'onlineSavingsMsg'
Learn More

Yttrium foil, 0.1mm (0.004in) thick, 99.6% (REO), Thermo Scientific Chemicals

127.00 € - 407.00 €

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371799
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012442.FF
25 x 25 mm
127.00 €
25mm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11490280
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012442.FI
50 x 50 mm
407.00 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-65-5
88.91
VWQVUPCCIRVNHF-UHFFFAOYSA-N
23993
yttrium
Y
MFCD00011468
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
CHEBI:33331
[Y]

Specifications

7440-65-5
MFCD00011468
VWQVUPCCIRVNHF-UHFFFAOYSA-N
yttrium
23993
99.9%
Mylar under Argon
Foil
Yttrium foil, 0.1mm (0.004 in.) thick
Y
yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
[Y]
88.91
CHEBI:33331
Odorless
≈0.28g/25x25mm
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-174-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only