missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

36.88 € - 526.62 €

Chemical Identifiers

CAS 13494-98-9
Molecular Formula H12N3O15Y
Molecular Weight (g/mol) 383.008
MDL Number MFCD00149940
InChI Key QBAZWXKSCUESGU-UHFFFAOYSA-N
Synonym yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
PubChem CID 166833
IUPAC Name yttrium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327637
SDS View Documents
Thermo Scientific Alfa Aesar
012898.06
5 g
36.88 €
5g
Please to purchase this item. Need a web account? Register with us today!
11317637
SDS View Documents
Thermo Scientific Alfa Aesar
012898.14
25 g
52.86 €
25g
Please to purchase this item. Need a web account? Register with us today!
11337637
SDS View Documents
Thermo Scientific Alfa Aesar
012898.22
100 g
147.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
15426005
SDS View Documents
Thermo Scientific Alfa Aesar
012898.59
5 x 100 g
526.62 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13494-98-9
383.008
QBAZWXKSCUESGU-UHFFFAOYSA-N
166833
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]
H12N3O15Y
MFCD00149940
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
yttrium(3+);trinitrate;hexahydrate

Specifications

13494-98-9
99.9% (REO)
MFCD00149940
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]
383.008
383.01 (274.92 Anhydrous)
Yttrium(III) nitrate hexahydrate
Crystalline
H12N3O15Y
yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
QBAZWXKSCUESGU-UHFFFAOYSA-N
yttrium(3+);trinitrate;hexahydrate
166833
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 233-802-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.