missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) sulfide, 99.9% (REO), Thermo Scientific Chemicals

62.21 € - 1107.51 €

Chemical Identifiers

CAS 12039-19-9
Molecular Formula S3Y2
Molecular Weight (g/mol) 273.99
MDL Number MFCD00058806
InChI Key BPMRLDMEAPSVQN-UHFFFAOYSA-N
Synonym yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g
PubChem CID 166021
SMILES [S--].[S--].[S--].[Y+3].[Y+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11378378
View Documents
Thermo Scientific Alfa Aesar
039565.03
1 g
62.21 €
1g
Please to purchase this item. Need a web account? Register with us today!
11388378
View Documents
Thermo Scientific Alfa Aesar
039565.06
5 g
302.51 €
5g
Please to purchase this item. Need a web account? Register with us today!
11450231
View Documents
Thermo Scientific Alfa Aesar
039565.14
25 g
1107.51 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12039-19-9
273.99
BPMRLDMEAPSVQN-UHFFFAOYSA-N
166021
S3Y2
MFCD00058806
yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g
[S--].[S--].[S--].[Y+3].[Y+3]

Specifications

12039-19-9
S3Y2
UN3134
Soluble in water.
[S--].[S--].[S--].[Y+3].[Y+3]
273.99
273.93
Like rotten eggs
Yttrium(III) sulfide
Powder
MFCD00058806
yttrium sulfide, diyttrium trisulphide, yttrium iii sulfide, yttrium sulfide y2s3, yttrium 3+ trisulfide, diyttrium 3+ ion trisulfanediide, yttrium iii sulfide trace rare earth metals basis 1g
BPMRLDMEAPSVQN-UHFFFAOYSA-N
diyttrium(3+) trisulfanediide
166021
99.9%
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P335+P334-P362-P370+P378q-P501c

H261-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 234-887-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.